Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Tube Washer
Product Code. 15348638
Change view
Click to view available options
Length (Metric):
178 mm
203 mm
Diameter (Metric) Outer:
10 mm Tube Diameter, 14 mm B Diameter
5 mm Tube Diameter, 10 mm B Diameter
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Outer Unit Size
15348638 178 mm 5 mm Tube Diameter, 10 mm B Diameter Each
15328638 178 mm 10 mm Tube Diameter, 14 mm B Diameter Each
15358638 203 mm 5 mm Tube Diameter, 10 mm B Diameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15348638 Supplier Bel-Art™ Supplier No. 660000064
  • ONLINE EXCLUSIVE PRICE
    228.00€
    211.01€ / 1 pièces
    Économisez 16.99€
    7% de réduction
View all promotions
Expédition estimée: 15-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Wiorks with filter flask and inverted sample tube. Bel-Art SP Scienceware™ Wilmad™ Tube Washer with simple design washing unit is the most economical and reliable for single tube cleaning.
  • Each unit only accommodates NMR tube with OD and length as indicated in specifications
  • Requires filter flask (sold separately; 500ml recommended)
  • Inverted sample tube is inserted into the washer and solvent is introduced into the reservoir using a series of wash bottles.
  • Pulls air through the tube to complete the procedure.
Specifications
Length (Metric) 178 mm
Material Borosilicate Glass
Product Type NMR Tube
Diameter (Metric) Outer 5 mm Tube Diameter, 10 mm B Diameter
Diameter (Metric) Inner 24/40 inner joint
Length (English) 7 in.