Learn More
Hirschmann™ Glass Inserts
Product Code. 30158520
Change view
Click to view available options
No. per Pack:
384/Pk.
Unit Size:
Pack of 19200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
30158520 384/Pk. Pack of 19200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30158520 Supplier Hirschmann™ Supplier No. 9240584
  • ONLINE EXCLUSIVE PRICE
    9245.00€
    5600.00€ / 19200 pièces
    Économisez 3645.00€
    39% de réduction
Expédition estimée: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Glass inserts for Hirschmann™ plates formats. Glass providing excellent chemical inertness, no leaching of plastics.
  • Loose glass inserts
  • Highly pure
  • Chemically inert
Specifications
Material DURAN™ Glass
No. per Pack 384/Pk.
For Use With (Equipment) 384-well Hirschmann plates
Product Type Glass Inserts