Learn More
XRP44X, TRC
Product Code. 30450031
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450031 100 mg 100mg
30448948 10 mg 10mg
30456506 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450031 Supplier Toronto Research Chemicals Supplier No. TRCX750960100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 729605-21-4
Molecular Formula C21H21ClN4O
Molecular Weight (g/mol) 380.87
IUPAC Name [4-(3-chlorophenyl)piperazin-1-yl]-(5-methyl-2-phenylpyrazol-3-yl)methanone
SMILES Cc1cc(C(=O)N2CCN(CC2)c3cccc(Cl)c3)n(n1)c4ccccc4
Specifications
Quantity 100 mg
Synonym XRP44X, 4-(3-Chlorophenyl)-1-piperazinyl](3-methyl-1-phenyl-1H-pyrazol-5-yl)methanone, XRP 44X, [4-(3-Chlorophenyl)piperazin-1-yl](5-methyl-2-phenyl-2H-pyrazol-3-yl)methanone, [4-(3-Chlorophenyl)piperazin-1-yl]-(5-methyl-2-phenylpyrazol-3-yl)methanone
InChI Formula InChI=1S/C21H21ClN4O/c1-16-14-20(26(23-16)18-7-3-2-4-8-18)21(27)25-12-10-24(11-13-25)19-9-5-6-17(22)15-19/h2-9,14-15H,10-13H2,1H3
Formula Weight 380.1404
Chemical Name or Material 1-(3-Chlorophenyl)-4-[(3-methyl-1-phenyl-1H-pyrazol-5-yl)carbonyl]piperazine