Learn More
Abcam XTT Sodium Salt, Tetrazolium Salt, Solid, 25 mg
Product Code. 32120928 Shop All Abcam Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120928 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120928 Supplier Abcam Supplier No. ab14631025mg
 Restricted Item
Expédition estimée: 24-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Tetrazolium salt for colorimetric cell viability and proliferation assays.
  • Molecular weight: 673.53 g/mol
  • Form: Solid
  • Applications: Used in colorimetric assays for cell viability and proliferation. Cleavage of XTT by dehydrogenase enzymes of metabolically active cells yields a highly colored, water-soluble formazan product.
  • Advantages: Does not require a formazan crystal solubilization step unlike MTT assays.
  • Usage examples: Has been used to assess valvular injury for testing cryopreserved cardiac valves.
The product is subject to the following: Abcam Restricted Use Statement
Spezifikation
Content And Storage Store at -20°C.
Detection Location In-Solution Detection
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate Dehydrogenase Enzyme Substrate
Target Molecule Dehydrogenase Enzymes
Detection Method Colorimetric
Quantity 25 mg
For Use With (Application) Colorimetric assays for cell viability and proliferation, valvular injury assessment for cryopreserved cardiac valves
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.