Learn More
Thermo Scientific Chemicals Yeast Extract
Product Code. 15493079
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493079 1 kg 1kg
15483079 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493079 Supplier Thermo Scientific Chemicals Supplier No. J60287.A1
  • ONLINE EXCLUSIVE PRICE
    198.90€
    164.65€ / 1kg
    Économisez 34.25€
    17% de réduction
View all promotions
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy
Used as food additive, flavor enhancer in processed foods of all kinds and as nutrient for bacterial culture media. It is used to create savoury flavors and umami taste sensations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
TRUSTED_SUSTAINABILITY
Spezifikation
Form Powder
Packaging Type Plastic Bottle
Quantity 1 kg
RUO – Research Use Only
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.