Learn More
YSI™ Membrane kits
Product Code. 11900095
Change view
Click to view available options
Probe Type:
DO
Oxygen/Temperature
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Probe Type Unit Size
11900095 DO Each
11984366 Oxygen/Temperature Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11900095 Supplier YSI™ Supplier No. 605306
  • ONLINE EXCLUSIVE PRICE
    78.20€
    75.59€ / 1 pièces
    Économisez 2.61€
    3% de réduction
 Restricted Item
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
For Use With (Equipment) YSI™ Model 556
Type Membrane Kit
Includes Six 1.25 mil. cap membranes, KCl solution
Probe Type DO