Learn More
(Z)-11-Hexadecenal, TRC
Product Code. 30407705
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407705 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407705 Supplier Toronto Research Chemicals Supplier No. TRCH293700100MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (Z)-11-Hexadecenal
CAS 53939-28-9
Molecular Formula C16 H30 O
Quantity 100 mg
Synonym 11-Hexadecenal, (Z)-, (11Z)-11-Hexadecenal, (Z)-11-Hexadecen-1-al, (Z)-11-Hexadecenal, (Z)-11-Hexadecenyl aldehyde, 11(Z)-Hexadecenal, cis-11-Hexadecen-1-al, cis-11-Hexadecenal
InChI Formula InChI=1S/C16H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h5-6,16H,2-4,7-15H2,1H3/b6-5-
SMILES CCCC\C=C/CCCCCCCCCC=O
IUPAC Name (Z)-hexadec-11-enal
Molecular Weight (g/mol) 238.41
Formula Weight 238.2297