Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 17 Peptide
Product Code. 10328882
Change view
Click to view available options
Quantity:
800 x 20μL, 800 x 20 μL Assays
Unit Size:
16000µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10328882 800 x 20μL, 800 x 20 μL Assays 16000µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10328882 Supplier Thermo Scientific™ Supplier No. PV4316
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon glycogen synthase, containing Ser-7.
TRUSTED_SUSTAINABILITY
Specifications
Description Z'-LYTE™ Ser/Thr 17 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Serine/Threonine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Serine/Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis
Product Line Z ́-LYTE
For Use With (Application) Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acute-lyte™ kinase assays, enzyme and protein activity assays, protein assays and, FRET
Includes Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer
Sample Volume 800 x 20μL
Content And Storage –80°C, Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
Assay Serine/Threonine Kinase Assay
Assay Entry Biochemical phospho-specific activity (no antibody)
Product Type Z'-LYTE™ Ser/Thr 17 Peptide Kinase Assay Kit
Quantity 800 x 20μL, 800 x 20 μL Assays
Format 384-well plate
Excitation/Emission 405/465, 535 nm
Label or Dye Methylcoumarin
No. of Reactions 800 Reactions
Readout End Point
References Biochem J. vol 296, pp. 843-9 (1993)
Shipping Condition Dry Ice
Source Similar to Ser-7 of Glycogen Synthase KKLNRTLSVA (KKLNRTLSFAEPG)
Substrate Properties Peptide-Based Substrate
Target Entry CAMK2B, PRKCN, PRKD2, DCAMKL2, PRKD1, NEK4, MELK
Target Enzyme Serine/Threonine Kinases
Validated Kinases CAMK2B, DCAMKL2 , MELK, NEK4, PRKCN , PRKD1, PRKD2
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.