Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 7 Peptide
Product Code. 10492902
Change view
Click to view available options
Quantity:
800 x 20μL, 800 x 20 μL Assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10492902 800 x 20μL, 800 x 20 μL Assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10492902 Supplier Thermo Scientific™ Supplier No. PV4305
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon LAT, containing Tyr-200.
TRUSTED_SUSTAINABILITY
Specifications
Description Z'-LYTE™ Tyrosine 7 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Liquid, Tyrosine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis
Product Line Z ́-LYTE
Formulation Liquid
For Use With (Application) Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acutelyte™ kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity, FRET
Includes Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer
Sample Volume 800 x 20μL
Assay Tyrosine Kinase Assay
Assay Entry Biochemical phospho-specific activity (no antibody)
Product Type Z'-LYTE™ Tyrosine 7 Peptide Kinase Assay Kit
Quantity 800 x 20μL, 800 x 20 μL Assays
Format 384-well plate
Content And Storage Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. See COA for specific storage instructions Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
Excitation/Emission 405/465, 535 nm
Label or Dye Methylcoumarin
No. of Reactions 800
Readout End Point
References Biochem. J. 356 p 461-71 (2001)
Shipping Condition Dry Ice
Source Based on LAT, Residues 195 to 208, Centering Y-200 (ESIDDYVNVPESGES)
Substrate Properties Peptide-Based Substrate
Target Entry ZAP70
Target Enzyme Tyrosine Kinases
Validated Kinases ZAP70
Show More Show Less
For Research Use Only. Not for use in diagnostic procedures.