missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TerraCycle™ Zero Waste Box™ Disposable Gloves Recycling Solution, 5-pack Bundle, France
Description
- Reduce the amount of waste that sent to landfill or incineration.
- Pack of 5 Large 140 L recyclable Zero Waste Boxes to recycle vinyl, nitrile, and latex gloves.
- Please do not include gloves that are contaminated with materials considered hazardous or dangerous.
- DO NOT RECYCLE:
- Other types of PPE, medical waste (this includes material contaminated with blood, bodily fluids and waste which should be collected in a hazardous clinical waste bin)
- Autoclaved material
- Heavy-duty, fabric, and leather gloves
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious, or pathogenic).
- Each box is supplied with a pre-affixed return shipping label.
- How it works:
- Order 5-pack bundle.
- Collect: Fill the boxes with the accepted items.
- Ship: Once full, use the pre-paid shipping label (already fastened to the boxes) to arrange for waste to be collected for recycling.
- Reorder: Order your next Zero Waste Boxes to continue to recycle.
- Everything collected is sorted and processed into raw materials that can be reused instead of being sent to landfill or incineration.
- The boxes are made in France from 100% recycled material.
Specifications
Specifications
| Capacity (Metric) | 25 kg |
| Certifications/Compliance | FSC |
| Capacity (English) | 140 L |
| Color | Brown, Green |
| Dimensions (L x W x H) | 38 x 38 x 97 cm |
| Disposable | Single-use |
| Height (Metric) | 97 cm |
| Latex Free | Yes |
| Length (Metric) | 38 cm |
| Sterility | Non-sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?