missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Zero Waste Box™ Rigid Lab Plastics Recycling Solution, 5-pack Bundle, France

Product Code. 32096794
Change view
Click to view available options
Capacity (Metric):
25 kg
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
32096794 25 kg Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32096794 Supplier TerraCycle™ Supplier No. CS129LFR_BUNDLE
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support sustainability goals with this easy-to-use recycling solution for laboratory plastics. Zero waste boxes are an all-inclusive eco-friendly waste disposal service which includes storage, shipping, and processing of the materials.

  • Reduce the amount of waste sent to landfill or incineration.
  • Pack of 5 Large 140 L recyclable Zero Waste Boxes.
  • Ideal for any brand and type of rigid lab plastics such as centrifuge tubes, plastic bottles, trays, vials, beakers, plastic spatulas, and pipette tips.
  • DO NOT RECYCLE
    • Tubes and rigid lab plastics that are contaminated with bodily fluids and materials considered hazardous or dangerous.
    • Autoclaved material.
    • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious, or pathogenic)
    • Batteries, pressurised canisters, broken glass and medical waste.
  • Each box is supplied with a pre-affixed return shipping label.
  • How it works:
    • Order your 5-pack bundle.
    • Collect: Fill the boxes with the accepted items.
    • Ship: Once full, use the pre-paid shipping label (already fastened to the boxes) to arrange for your waste to be collected for recycling.
    • Reorder: Order your next Zero Waste Boxes to continue to recycle.
  • Everything collected is sorted and processed into raw materials that can be reused instead of being sent to landfill or incineration.
  • The boxes are made in France from 100% recycled material.

Specifications

Capacity (Metric) 25 kg
Certifications/Compliance FSC
Capacity (English) 140 L
Color Brown, Green
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Height (Metric) 97 cm
Latex Free Yes
Length (Metric) 38 cm
Sterility Non-sterile
Width (Metric) 38 cm
Autoclavable Not Autoclavable
Includes Return Shipping Label
For Use With (Application) Rigid Lab Plastics Recycling Waste
Packaging France
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.