missing translation for 'onlineSavingsMsg'
Learn More

Zinc propionate, 97%

Product Code. 11480028
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480028 1000 g 1000g
11490028 50 g 50g
11400038 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480028 Supplier Thermo Scientific Alfa Aesar Supplier No. B23636.0B
 Restricted Item
Expédition estimée: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 557-28-8 | C6H10O4Zn | 211.52 g/mol

Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.

Solubility
Soluble in alcohol, ether, chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.

Notes
Hygroscopic, protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-28-8
Molecular Formula C6H10O4Zn
Molecular Weight (g/mol) 211.52
MDL Number MFCD00045937
InChI Key XDWXRAYGALQIFG-UHFFFAOYSA-L
Synonym zinc propionate, unii-81pxa87xvc, 81pxa87xvc, propanoic acid, zinc salt 2:1, gyna-plex, propionic acid, zinc salt, zinc dipropionate, propanoic acid, zinc salt, zinc propanoate, propanoic acid, zinc salt, basic
PubChem CID 11189
IUPAC Name zinc;propanoate
SMILES [Zn++].CCC([O-])=O.CCC([O-])=O

Specifications

Quantity 1000 g
Sensitivity Hygroscopic
Merck Index 14,10153
Solubility Information Soluble in alcohol,ether,chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.
Formula Weight 211.53
Percent Purity 97%
Chemical Name or Material Zinc propionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.