CAS RN 124219-00-7
CAS RN 124219-00-7
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
MedChemExpress Asp-Asp-Asp-Asp-Asp
MedChemExpress Asp-Asp-Asp-Asp-Asp is a peptide consists of 5 Asp.
2
MedChemExpress Asp-Asp-Asp-Asp-Asp
MedChemExpress Asp-Asp-Asp-Asp-Asp is a peptide consists of 5 Asp.
3
MedChemExpress Asp-Asp-Asp-Asp-Asp
MedChemExpress Asp-Asp-Asp-Asp-Asp is a peptide consists of 5 Asp.