CAS RN 125911-68-4
CAS RN 125911-68-4
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
MedChemExpress SAMS
MedChemExpress SAMS peptide is a specific substrate for the AMP-activated protein kinase (AMPK).
2
MedChemExpress SAMS
MedChemExpress SAMS peptide is a specific substrate for the AMP-activated protein kinase (AMPK).
3
MedChemExpress SAMS
MedChemExpress SAMS peptide is a specific substrate for the AMP-activated protein kinase (AMPK).