CAS RN 1449236-96-7
CAS RN 1449236-96-7
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
NMS-859, MedChemExpress
MedChemExpress NMS-859 is a potent, covalent VCP (p97) inhibitor, with IC50s of 0.37 and 0.36 μM for wild-type VCP in the presence of 60 μM and 1 mM ATP in cells, respectively.