CAS RN 1800025-30-2
CAS RN 1800025-30-2
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
MGAT2-IN-1, MedChemExpress
MedChemExpress MGAT2-IN-1 is an orally active inhibitor of monoacylglycerol acyltransferase (MGAT2) with IC50 of 7.8 and 2.4 nM for human and mouse MGAT2, respectively.