CAS RN 309-00-2
CAS RN 309-00-2
IUPAC Nor:
1,8,9,10,11,11-hexachlorotetracyclo[6.2.1.1³,⁶.0²,⁷]dodeca-4,9-diene
Synonymes:
aldrin (iso)
Poids moléculaire (g/mol):
364.9
Formule moléculaire:
C12H8Cl6
InChi Key:
QBYJBZPUGVGKQQ-UHFFFAOYNA-N
SMILES:
ClC1=C(Cl)C2(Cl)C3C4CC(C=C4)C3C1(Cl)C2(Cl)Cl
1
–
6
de
6
résultats
3
Aldrin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Aldrin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Aldrin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.