CAS RN 3100-36-5
CAS RN 3100-36-5
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
Animusk (~65%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Animusk (~65%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Animusk (~65%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.