CAS RN 31264-51-4
CAS RN 31264-51-4
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
3-(2,5-Dimethylphenoxy)propyl Chloride (3-(2,5-Xylyloxy)propyl Chloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.