CAS RN 315679-65-3
CAS RN 315679-65-3
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
2-Amino-4-[4-(phenylmethyl)phenyl]-3-thiophenecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Amino-4-[4-(phenylmethyl)phenyl]-3-thiophenecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Amino-4-[4-(phenylmethyl)phenyl]-3-thiophenecarboxylic Acid Ethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.