CAS RN 31570-04-4
CAS RN 31570-04-4
IUPAC Nor:
tris(2,4-di-tert-butylphenyl) phosphite
Synonymes:
tris(2,4-di-tert-butylphenyl) phosphite
Poids moléculaire (g/mol):
646.94
Formule moléculaire:
C42H63O3P
InChi Key:
JKIJEFPNVSHHEI-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(OP(OC2=CC=C(C=C2C(C)(C)C)C(C)(C)C)OC2=CC=C(C=C2C(C)(C)C)C(C)(C)C)C(=C1)C(C)(C)C
1
–
2
de
2
résultats
1
Tris(2,4-di-tert-butylphenyl)phosphite, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Tris(2,4-di-tert-butylphenyl)phosphite 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality