CAS RN 31581-32-5
CAS RN 31581-32-5
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
Ethyl Atrarate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.