CAS RN 317323-77-6
CAS RN 317323-77-6
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
(3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol 3-(4-Methylbenzenesulfonate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.