CAS RN 331244-89-4
CAS RN 331244-89-4
IUPAC Nor:
4-[2-(2-ethoxyphenyl)hydrazin-1-ylidene]-3-methyl-1-(4-phenyl-1,3-thiazol-2-yl)-4,5-dihydro-1H-pyrazol-5-one
Synonymes:
4-[2-(2-ethoxyphenyl)hydrazin-1-ylidene]-5-methyl-2-(4-phenyl-1,3-thiazol-2-yl)pyrazol-3-one
Poids moléculaire (g/mol):
405.48
Formule moléculaire:
C21H19N5O2S
InChi Key:
KOCVKGYKBLJEPK-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=C1NN=C1C(C)=NN(C1=O)C1=NC(=CS1)C1=CC=CC=C1
1
–
1
de
1
résultats
1
BAM7, MedChemExpress
MedChemExpress BAM7 is a direct and selective activator of proapoptotic BAX with an IC50 of 3.3 μM.