CAS RN 481658-94-0
CAS RN 481658-94-0
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
Dirlotapide, MedChemExpress
MedChemExpress Dirlotapide (CP742033) is a gut-selective inhibitor of microsomal triglyceride transfer protein (MTP) that reliably produces weight loss in obese dogs.