CAS RN 58100-03-1
CAS RN 58100-03-1
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
MedChemExpress Tyr-Somatostatin-14
MedChemExpress Tyr-Somatostatin-14 is a customized peptide that adds a Tyrosine amino acid to Somatostatin-14.
2
MedChemExpress Tyr-Somatostatin-14
MedChemExpress Tyr-Somatostatin-14 is a customized peptide that adds a Tyrosine amino acid to Somatostatin-14.
3
MedChemExpress Tyr-Somatostatin-14
MedChemExpress Tyr-Somatostatin-14 is a customized peptide that adds a Tyrosine amino acid to Somatostatin-14.