CAS RN 845272-21-1
CAS RN 845272-21-1
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
Varlitinib, MedChemExpress
MedChemExpress Varlitinib (ASLAN001) is a potent, reversible, small molecule pan-EGFR inhibitor with IC50s of 7, 2, 4 nM for HER1, HER2 and HER4, respectively.