CAS RN 882663-88-9
CAS RN 882663-88-9
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
résultats
1
AMG-47a, MedChemExpress
MedChemExpress AMG-47a is a potent and orally active lymphocyte-specific protein tyrosine kinase (Lck) inhibitor, with an IC50 of 0.2 nM. AMG-47a also inhibits VEGF2, p38α, Jak3 and MLR and IL-2 with IC50s of 1 nM, 3 nM, 72 nM, 30 nM and 21 nM, respectively. AMG-47a has anti-inflammatory activity.