CAS RN 90419-12-8
CAS RN 90419-12-8
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
résultats
1
MedChemExpress Pancreatic Polypeptide, rat
MedChemExpress Pancreatic Polypeptide, rat is an agonist of NPY receptor, with high affinity at NPYR4.
2
MedChemExpress Pancreatic Polypeptide, rat
MedChemExpress Pancreatic Polypeptide, rat is an agonist of NPY receptor, with high affinity at NPYR4.