CAS RN 983-79-9
CAS RN 983-79-9
IUPAC Nor:
bis(diphenylmethylidene)hydrazine
Synonymes:
bis(diphenylmethylidene)hydrazine
Poids moléculaire (g/mol):
360.46
Formule moléculaire:
C26H20N2
InChi Key:
VQSIVLYYQQCXAF-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C(=NN=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1