CAS RN 117928-93-5
CAS RN 117928-93-5
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
résultats
1
MedChemExpress NT 13
MedChemExpress NT 13 (TPPT) is a tetrapeptide having the amino acid sequence L-threonyl-L-prolyl-L-prolyl-L-threonine amide. NT 13 is a partial N-methyl-D-aspartate receptor (NMDAR) agonist used in the study of depression, anxiety, and other related diseases.
2
MedChemExpress NT 13
MedChemExpress NT 13 (TPPT) is a tetrapeptide having the amino acid sequence L-threonyl-L-prolyl-L-prolyl-L-threonine amide. NT 13 is a partial N-methyl-D-aspartate receptor (NMDAR) agonist used in the study of depression, anxiety, and other related diseases.