CAS RN 124454-83-7
CAS RN 124454-83-7
IUPAC Nor:
—
Synonymes:
—
Poids moléculaire (g/mol):
—
Formule moléculaire:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
résultats
1
MedChemExpress Nucleoprotein (118-126)
MedChemExpress Nucleoprotein (118-126) is a 9-aa peptide, a fragment of Nucleoprotein.
2
MedChemExpress Nucleoprotein (118-126)
MedChemExpress Nucleoprotein (118-126) is a 9-aa peptide, a fragment of Nucleoprotein.
3
MedChemExpress Nucleoprotein (118-126)
MedChemExpress Nucleoprotein (118-126) is a 9-aa peptide, a fragment of Nucleoprotein.