CAS RN 1309980-11-7
CAS RN 1309980-11-7
IUPAC Nor:
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
Synonymes:
2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propan-2-ol
Poids moléculaire (g/mol):
262.16
Formule moléculaire:
C15H23BO3
InChi Key:
NNPQWBXXZHNLAW-UHFFFAOYSA-N
SMILES:
CC(C)(O)C1=CC=CC(=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
de
1
résultats
1
Ester pinacolique d’acide 3-(2-hydroxy-2-propanyl)phénylboronique, 97 %, ACROS Organics™
CAS: 1309980-11-7 Clé InChI: NNPQWBXXZHNLAW-UHFFFAOYSA-N Synonyme: 2-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol,3-2-hydroxypropan-2-yl phenylboronic acid pinacol ester,3-2-hydroxy-2-propyl phenylboronic acid pinacol ester,3-2-hydroxy-2-propanyl phenylboronic acid pinacol ester,3-2-hydroxypropan-2-yl phenyl boronic acid pinacol ester,2-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl propan-2-ol,2-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl propan-2-ol CID PubChem: 46738028 Nom IUPAC: 2-[3-(4,4,5,5-tétraméthyl-1,3,2-dioxaborolan-2-yl)phényl]propan-2-ol SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(C)(C)O