missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-thien-2-yl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10122794
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10122794 1 g Amber Glass Bottle 1g
11380647 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10122794 Supplier Thermo Scientific Maybridge Supplier No. CC47201DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 871825-56-8
Molecular Formula C9H8N2O2S
Molecular Weight (g/mol) 208.24
MDL Number MFCD08271954
InChI Key YSTOGMCNCZGBTG-UHFFFAOYSA-N
Synonym 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxylic acid, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-thiophen-2-yl-2h-pyrazole-3-carboxylic acid, 2-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1-methyl-3-2-thienyl-1 h-pyrazole-5-carboxylic acid, 1-methyl-3-2-thienyl pyrazole-5-carboxylic acid, 2-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 5-carboxy-1-methyl-3-thien-2-yl-1h-pyrazole, 2-5-carboxy-1-methyl-1h-pyrazol-3-yl thiophene
PubChem CID 20109897
IUPAC Name 2-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid
SMILES CN1N=C(C=C1C(O)=O)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-thien-2-yl-1H-pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.