Learn More
Viking Optical™ 10X Folding Magnifier
Code produit 12886325
Encompass_Preferred
Modifier l'affichage
Click to view available options
Diameter (Metric) Lens:
19 mm
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Diameter (Metric) Lens Conditionnement
12886325 19 mm 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 12886325 Fournisseur Viking Optical™ Code fournisseur 5609
Expédition estimée: 21-10-2026
Pocket doublet type, giving tolerably flat field with minimum chromatic aberration. Lens approximately 19mm diameter. Mounted in metal frame.
Spécification
Diameter (Metric) Lens 19 mm