Learn More
17-Desethynyl Norethindrone Diacetate, TRC
Code produit 30361068
Change view
Click to view available options
Quantité:
100 mg
1 g
Conditionnement:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30361068 1 g 1g
30354783 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30361068 Fournisseur Toronto Research Chemicals Code fournisseur TRCD2897651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 66964-58-7
Formule moléculaire C22 H30 O4
Poids moléculaire (g/mol) 358.47
Synonyme 19-Norpregn-4-ene-3,20-dione, 17-(acetyloxy)-, (17α)- (9CI), 19-Nor-17α-pregn-4-ene-3,20-dione, 17-hydroxy-, acetate (6CI,7CI), 3,20-Dioxo-19-nor-17α-pregn-4-en-17-yl acetate, 17-Desethynyl Norethindrone Diacetate, Norethisterone Acetate Imp. E (EP)
Nom IUPAC [(8R,9S,10R,13S,14S,17S)-17-acetyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C(=O)C
Spécification
Nom chimique ou matériau 17-Desethynyl Norethindrone Diacetate
Quantité 1 g
Formule InChI InChI=1S/C22H30O4/c1-13(23)22(26-14(2)24)11-9-20-19-6-4-15-12-16(25)5-7-17(15)18(19)8-10-21(20,22)3/h12,17-20H,4-11H2,1-3H3/t17-,18+,19+,20-,21-,22+/m0/s1
Poids de la formule 358.2144