Learn More
17β-Estradiol-2,4,16,16,17-d5, TRC
Code produit 30376887
Change view
Click to view available options
Quantité:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30376887 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30376887 Supplier Toronto Research Chemicals Supplier No. TRCE8880065MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Molécules organiques de haute pureté et étalons analytiques, distribués stratégiquement dans le monde entier pour favoriser l'innovation et la réussite commerciale.
TRUSTED_SUSTAINABILITY
Specifications
Nom chimique ou matériau 17beta-Estradiol-D5
CAS 221093-45-4
Notes de qualité de la pureté HPLC
Formule moléculaire C18 2H5 H19 O2
Quantité 5 mg
Synonyme Estra-1,3,5(10)-triene-2,4,16,16,17-d5-3,17-diol, (17β)-, (17β)-Estra-1,3,5(10)-triene-2,4,16,16,17-d5-3,17-diol, 17β-Estradiol-D5, 17β-Estradiol-2,4,16,16,17-D5
Formule InChI InChI=1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1/i3D,7D2,10D,17D
SMILES [2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])[C@]4([2H])O)[C@@H]3CCc2c([2H])c1O
Nom IUPAC (8R,9S,13S,14S,17S)-2,4,16,16,17-pentadeuterio-13-metil-6,7,8,9,11,12,14,15-octahidrociclopenta[a]fenantreno-3,17-diol
Poids moléculaire (g/mol) 277.41
Poids de la formule 277.209
Pourcentage de pureté >95