Learn More
R&D Systems™ 1A-116
Code produit 18054522
Modifier l'affichage
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18054522 10 mg 10mg
18769846 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18054522 Fournisseur R&D Systems™ Code fournisseur 6701/10
 Article restreint
Expédition estimée: 27-11-2026
A Tocris product, part of R&D Systems. Rac1 inhibitor; blocks Rac1-P-Rex1 interaction
1A-116 is a Rac1 inhibitor. Reduces intracellular Rac1-GTP levels and blocks Rac1-P-Rex1 interaction in vitro. Exhibits no effect on GTPase activity of closely related Cdc42. Displays antiproliferative, anti-invasive and pro-apoptotic activity in several cancer cell lines. Exhibits antimetastatic activity in vivo. Also restores antiproliferative effects of Tamoxifen (Cat. No. 6342) in resistant breast cancer cell.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau N-(3,5-Dimethylphenyl)-N-[2-(trifluoromethyl)phenyl]guanidine
CAS 1430208-73-3
Quantité 10 mg
Gamme de produits Tocris
Formule moléculaire C16H16F3N3
Poids de la formule 307.31
Type de produit Inhibitor
Pureté 98%