Learn More
2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol Dihydrochloride, TRC
Code produit 30369457
Change view
Click to view available options
Quantité:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30369457 50 mg
30382752 10 mg
30378874 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30369457 Fournisseur Toronto Research Chemicals Code fournisseur TRCD41868650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 1825392-15-1
Formule moléculaire C25H33N3O4S.2HCl
Poids moléculaire (g/mol) 544.53
Synonyme 2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol Dihydrochloride, Ethanol, 2-[2-[2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethoxy]ethoxy]-, hydrochloride (1:2) (ACI)
Nom IUPAC 2-[2-[2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethoxy]ethoxy]ethanol;dihydrochloride
SMILES Cl.Cl.OCCOCCOCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24
Spécification
Nom chimique ou matériau 2-[2-[2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethoxy]ethoxy]ethanol Dihydrochloride
Quantité 50 mg
Formule InChI InChI=1S/C25H33N3O4S.2ClH/c29-14-16-31-18-20-32-19-17-30-15-13-27-9-11-28(12-10-27)25-21-5-1-3-7-23(21)33-24-8-4-2-6-22(24)26-25;;/h1-8,29H,9-20H2;2*1H
Poids de la formule 543.1725