missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-4-pentenoic acid, 95%

Product Code. 15487438
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487438 5 g 5g
15477438 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487438 Supplier Thermo Scientific Alfa Aesar Supplier No. H60940.06
 Restricted Item
Expédition estimée: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 16386-93-9 | C7H12O2 | 128.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16386-93-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00012304
InChI Key BGUAPYRHJPWVEM-UHFFFAOYSA-N
Synonym 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g
PubChem CID 140065
IUPAC Name 2,2-dimethylpent-4-enoic acid
SMILES CC(C)(CC=C)C(O)=O

Specifications

Density 0.933
Boiling Point 104°C to 108°C
Flash Point 88°C (190°F)
Linear Formula H2C=CHCH2C(CH3)2CO2H
Refractive Index 1.431
Quantity 5 g
UN Number UN3265
Solubility Information Soluble in water.
Formula Weight 128.17
Percent Purity 95%
Chemical Name or Material 2,2-Dimethyl-4-pentenoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.