missing translation for 'onlineSavingsMsg'
Learn More

2,3-dihydro-1,4-benzodioxine-6-ylméthanol, Thermo Scientific™

Product Code. p-2696865
Change view
Click to view available options
Quantité:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité Unit Size
10262582 250 mg 250mg
10150582 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10262582 Supplier Thermo Scientific Maybridge Supplier No. CC01309CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 39270-39-8
Formule moléculaire C9H10O3
Poids moléculaire (g/mol) 166.18
Numéro MDL MFCD01317581
Clé InChI FFLHNBGNAWYMRH-UHFFFAOYSA-N
Synonyme 2,3-dihydrobenzo b 1,4 dioxin-6-yl methanol, 2,3-dihydro-1,4-benzodioxin-6-methanol, 1,4-benzodioxin-6-methanol, 2,3-dihydro, 1,4-benzodioxin-6-methanol,2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-6-yl methanol, 1,4-benzodioxan-6-methanol, 2,3-dihydro-1,4-benzodioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl methanol, 2,3-dihydrobenzo 1,4 dioxin-6-yl-methanol, 2,3-dihydro-benzo 1,4 dioxin-6-yl-methanol
CID PubChem 2776174
SMILES OCC1=CC=C2OCCOC2=C1

Specifications

Nom chimique ou matériau 2, 3-Dihydro-1, 4-benzodioxin-6-ylmethanol
% min. CAS 96.5
% max. CAS 100.0
Plage de pourcentage du dosage 97%
Quantité 250 mg
Nom IUPAC (2,3-dihydro-1,4-benzodioxin-6-yl)methanol
Poids de la formule 166.18
Pourcentage de pureté 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.