Learn More
2,4-Dichlorobenzyl alcohol, 99%
Description
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
Solubility
Soluble in water, methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 57°C to 61°C |
| Flash Point | >110°C (230°F) |
| Quantity | 100 g |
| Beilstein | 1448652 |
| Merck Index | 14,3060 |
| Solubility Information | Soluble in water,methanol and chloroform. |
| Formula Weight | 177.03 |
| Percent Purity | 99% |
| Chemical Name or Material | 2,4-Dichlorobenzyl alcohol |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.