missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyric acid, 98%

Product Code. 11418836
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418836 250 mL 250mL
11408836 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418836 Supplier Thermo Scientific Alfa Aesar Supplier No. A16989.AK
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy

CAS: 88-09-5 | C6H12O2 | 116.16 g/mol

Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetic acid (88-09-5) is used in flavoring agent and as an intermediate for pharmaceuticals and dyestuffs. It is also employed in the manufacture of esters and other chemicals.

Solubility
Sparingly soluble with the usual organic solvents. Solubility in water 18 g/L (20°C).

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-09-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002670
InChI Key OXQGTIUCKGYOAA-UHFFFAOYSA-N
Synonym 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid
PubChem CID 6915
IUPAC Name 2-ethylbutanoic acid
SMILES CCC(CC)C(=O)O

Specifications

Melting Point -15°C
Density 0.924
Boiling Point 192°C to 194°C
Flash Point 87°C (188°F)
Refractive Index 1.413
Quantity 250 mL
UN Number UN2922
Beilstein 1098634
Merck Index 14,3110
Solubility Information Sparingly soluble with the usual organic solvents. Solubility in water 18g/L (20°C).
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 2-Ethylbutyric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Ethylbutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.