Learn More
2-Formyl-cefpodoxime Proxetil, TRC
Code produit 30404884
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30404884 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30404884 Fournisseur Toronto Research Chemicals Code fournisseur TRCF69870010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 2-Formyl-cefpodoxime Proxetil
CAS 96680-30-7
Formule moléculaire C22H27N5O10S2
Quantité 10 mg
Synonyme (6R,7R)-7-[[(2Z)-2-[2-(Formylamino)-4-thiazolyl]-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 1-[[(1-methylethoxy)carbonyl]oxy]ethyl Ester, [6R-[6alpha,7beta(Z)]]-7-[[[2-(Formylamino)-4-thiazolyl](methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid, 1-[[(1-methylethoxy)carbonyl]oxy]ethyl Ester, 1-[[(1-Methylethoxy)carbonyl]oxy]ethyl, (6R,7R)-7-[[(2Z)-2-[2-(formylamino)-4-thiazolyl]-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefpodoxime Proxetil EP Impurity F, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-2-[2-(formylamino)-4-thiazolyl]-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-, 1-[[(1-methylethoxy)carbonyl]oxy]ethyl ester, (6R,7R)-
Formule InChI InChI=1S/C22H27N5O10S2/c1-10(2)35-22(32)37-11(3)36-20(31)16-12(6-33-4)7-38-19-15(18(30)27(16)19)25-17(29)14(26-34-5)13-8-39-21(24-13)23-9-28/h8-11,15,19H,6-7H2,1-5H3,(H,25,29)(H,23,24,28)/b26-14-/t11?,15-,19-/m1/s1
SMILES COCC1=C(N2[C@H](SC1)[C@H](NC(=O)\C(=N/OC)\c3csc(NC=O)n3)C2=O)C(=O)OC(C)OC(=O)OC(C)C
Poids moléculaire (g/mol) 585.61
Poids de la formule 585.1199