Learn More
2'-Hydroxybiphenyl-3-carboxylic Acid, TRC
Code produit 30406917
Change view
Click to view available options
Quantité:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30406917 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30406917 Fournisseur Toronto Research Chemicals Code fournisseur TRCH817605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 2'-Hydroxybiphenyl-3-carboxylic Acid
CAS 893736-72-6
Formule moléculaire C13H10O3
Quantité 100 mg
Synonyme 2'-Hydroxy[1,1'-biphenyl]-3-carboxylic acid
Formule InChI InChI=1S/C13H10O3/c14-12-7-2-1-6-11(12)9-4-3-5-10(8-9)13(15)16/h1-8,14H,(H,15,16)
SMILES OC(=O)c1cccc(c1)c2ccccc2O
Nom IUPAC 3-(2-hydroxyphenyl)benzoic acid
Poids moléculaire (g/mol) 214.22
Poids de la formule 214.063