Learn More
R&D Systems™ 2-Phospho-L-ascorbic acid trisodium salt
Description
2-Phospho-L-ascorbic acid trisodium salt is a stable ascorbic acid derivative used in cell culture; in combination with FGF-2, maintains differentiation potential in bone marrow-derived mesenchymal stem cells (MSC) through increased expression of HGF. Also exhibits synergistic protection of hMSCs under oxidative stress in combination with N-acetylcysteine (Cat. No. 5619).
Spécification
Spécification
| Nom chimique ou matériau | 2-(Dihydrogen phosphate)-L-ascorbic acid sodium salt |
| CAS | 66170-10-3 |
| Quantité | 50 mg |
| Gamme de produits | Tocris |
| Cible | Compounds for Stem Cell Differentiation |
| Formule moléculaire | C6H6Na3O9P |
| Poids de la formule | 322.05 |
| Pureté | >95% |