Learn More
Mecmesin™ Pince pour tissu 200 N
Code produit 11974088
Encompass
Modifier l'affichage
Click to view available options
Largeur (métrique):
100 mm
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Largeur (métrique) Conditionnement
11974088 100 mm 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11974088 Fournisseur Mecmesin™ Code fournisseur 432386
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
Fabric grips are designed for tensile and elongation testing of fabric and thin materials below 200N.
  • Material is gripped between two smooth spring-loaded faces which are clamped together by two locking nuts at either side of the grip to ensure the material is held securely across its entire width during testing.
  • Maximum specimen width: 100 mm
  • Maximum thickness: 4 mm.
  • Fitted with 10-32 UNF female hole and supplied with male grub-screw.
Spécification
Épaisseur 4 mm
Largeur (métrique) 100 mm