Learn More
3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxy-ethyl ester Benzenesulfonic acid, TRC
Code produit 30357511
Change view
Click to view available options
Quantité:
100 mg
Conditionnement:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30357511 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30357511 Fournisseur Toronto Research Chemicals Code fournisseur TRCD414610100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Nom chimique ou matériau 3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxy-ethyl ester Benzenesulfonic acid
CAS 1971915-89-5
Formule moléculaire C19H24N4O5S
Quantité 100 mg
Synonyme Benzenesulfonic acid, 3-(6,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxy-, ethyl ester
Formule InChI InChI=1S/C19H24N4O5S/c1-5-8-14-16-17(23(4)22-14)19(24)21-18(20-16)13-11-12(29(25,26)28-7-3)9-10-15(13)27-6-2/h9-11H,5-8H2,1-4H3,(H,20,21,24)
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)OCC
Nom IUPAC ethyl 4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonate
Poids moléculaire (g/mol) 420.48
Poids de la formule 420.1467