Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Spécification
| Nom chimique ou matériau | 3-Amino-6-chloropyridine |
| % min. CAS | 97.5 |
| % max. CAS | 100.0 |
| Point de fusion | 80°C to 84°C |
| Couleur | Marron |
| Spectre IR | Authentic |
| Plage de pourcentage du dosage | 98% |
| Conditionnement | Flacon en verre |
| Quantité | 1 g |
| Poids de la formule | 128.56 |
| Afficher plus |