Learn More
4-(3-Chloro-4-fluorophénylamino)-7-fluoro-6-nitroquinazoline, TRC
Product Code. 30354564
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantité:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
30354564 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30354564 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC37625010G
This item is not returnable. View return policy
Molécules organiques de haute pureté et étalons analytiques, distribués stratégiquement dans le monde entier pour favoriser l'innovation et la réussite commerciale.
TRUSTED_SUSTAINABILITY
Specifications
Nom chimique ou matériau 4-(3-Chloro-4-fluorophenylamino)-7-fluoro-6-nitroquinazoline
CAS 162012-67-1
Formule moléculaire C14H7ClF2N4O2
Quantité 10 g
Synonyme N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine, (3-Chloro-4-fluorophenyl)(7-fluoro-6-nitroquinazolin-4-yl)amine
Formule InChI InChI=1S/C14H7ClF2N4O2/c15-9-3-7(1-2-10(9)16)20-14-8-4-13(21(22)23)11(17)5-12(8)18-6-19-14/h1-6H,(H,18,19,20)
SMILES [O-][N+](=O)c1cc2c(Nc3ccc(F)c(Cl)c3)ncnc2cc1F
Nom IUPAC N-(3-chloro-4-fluorophényl)-7-fluoro-6-nitroquinazoline-4-amine
Poids moléculaire (g/mol) 336.68